Anti-GABRQ Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: Bioss
BS-12083R-FITC
BOSSBS-12083R-FITCEA 900 EUR
BOSSBS-12083R-FITC
Anti-GABRQ Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Gamma-aminobutyric acid type A (GABAA) receptors mediate inhibitory neurotransmission in the mammalian central nervous system. The receptor exists as a pentameric ion channel composed by heteromeric combinations of alpha, beta, gamma, delta, epsilon, theta, or pi subunits. Only specific subunit combinations produce viable receptors, while others never translocate to the cell surface from the ER where they are synthesized, and subsequently degraded. The theta subunit forms a receptor in combination with alpha3 subunits in monoaminergic cell groups. These receptors, found especially in theMore Product Information