Anti-PLPPR1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
BS-12150R-A680
BOSSBS-12150R-A680EA 890 EUR
BOSSBS-12150R-A680
Anti-PLPPR1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
This gene encodes a member of the plasticity-related gene (PRG) family. Members of the PRG family mediate lipid phosphate phosphatase activity in neurons and are known to be involved in neuronal plasticity. The protein encoded by this gene does not perform its function through enzymatic phospholipid degradation. This gene is strongly expressed in brain. It shows dynamic expression regulation during brain development and neuronal excitation. Alternatively spliced transcript variants encoding the same protein have been observed.

Type: Primary
Antigen: PLPPR1
Clonality: Polyclonal
Clone:
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR