Anti-NAV3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-12151R-HRP
BOSSBS-12151R-HRPEA 900 EUR
BOSSBS-12151R-HRP
Anti-NAV3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
May regulate IL2 production by T-cells. May be involved in neuron regeneration.

Recommended Dilutions: IHC-P: 1:100-500

Type: Primary
Antigen: NAV3
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR