Anti-Tryptophan 5-hydroxylase 1, Tryptophan 5-hydroxylase 2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
BS-1215R-A680
BOSSBS-1215R-A680EA 899 EUR
BOSSBS-1215R-A680
Anti-Tryptophan 5-hydroxylase 1, Tryptophan 5-hydroxylase 2 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
This gene encodes a member of the aromatic amino acid hydroxylase family. The encoded protein catalyses the first and rate limiting step in the biosynthesis of serotonin, an important hormone and neurotransmitter. Mutations in this gene have been associated with an elevated risk for a variety of diseases and disorders, including schizophrenia, somatic anxiety, anger-related traits, bipolar disorder, suicidal behavior, addictions, and others.

Type: Primary
Antigen: Tryptophan 5-hydroxylase 1, Tryptophan 5-hydroxylase 2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public ImmunogenMore Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR