Anti-SNX5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-12248R-A555
BOSSBS-12248R-A555EA 893 EUR
BOSSBS-12248R-A555
Anti-SNX5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Sorting nexin 1 (SNX1) is a member of a large family of hydrophilic proteins that interact with a variety of receptor types and are involved in intracellular trafficking (1). SNX1 and the related splice variant, SNX1A, bind the epidermal growth factor (EGF) receptor, facilitate its transport to lysosome, and thereby contribute to the degradation of the receptor (2,3). SNX2 and SNX4 share a high degree of amino acid similarity with SNX1, as they all contain a characteristic phox homology (PX) domain (4). These proteins are all partially associated with cellular membranes, and they, likewise,More Product Information
Order Now

SPECIFICATIONS

Antigen symbol SNX5
Environmentally Preferable
Antigen name Sorting Nexin 5
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 555
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 27131
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR