Anti-GPA33 Rabbit Polyclonal Antibody (Cy3®)
Supplier: Bioss
BS-1226R-CY3
BOSSBS-1226R-CY3EA 900 EUR
BOSSBS-1226R-CY3
Anti-GPA33 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
The glycoprotein encoded by this gene is a cell surface antigen that is expressed in greater than 95% of human colon cancers. The open reading frame encodes a 319 amino acid polypeptide having a putative secretory signal sequence and 3 potential glycosylation sites. The predicted mature protein has a 213 amino acid extracellular region, a single transmembrane domain, and a 62 amino acid intracellular tail. The sequence of the extracellular region contains 2 domains characteristic of the CD2 subgroup of the immunoglobulin (Ig) superfamily.
Recommended Dilutions: IF(IHC-P): 1:50-200