Anti-HOOK1 Rabbit Polyclonal Antibody (Cy3®)
Supplier: Bioss
BS-12287R-CY3
BOSSBS-12287R-CY3EA 883 EUR
BOSSBS-12287R-CY3
Anti-HOOK1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Microtubules mediate the spatial organization of diverse membrane-trafficking systems. The HOOK proteins, HOOK1, HOOK2 and HOOK3, comprise a family of cytosolic coiled-coil proteins that contain conserved N-terminal domains, which attach to microtubules; and more divergent C-terminal domains, which mediate binding to organelles. HOOK1, a cytoskeletal linker protein, may play a role in endocytic membrane trafficking. It exists as a homodimer, most likely mediated through its central coiled-coil domain. HOOK1 interacts with VPS18 and is required for spermatid differentiation, in which it isMore Product Information