Anti-LRRC19 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-12310R-A555
BOSSBS-12310R-A555EA 893 EUR
BOSSBS-12310R-A555
Anti-LRRC19 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
LRRC19

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LRRC19
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol LRRC19
Environmentally Preferable
Antigen name Leucine Rich Repeat Containing 19
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 555
Reactivity Human
Host Rabbit
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR