Anti-CRYGS Rabbit Polyclonal Antibody
Supplier: Bioss
BS-12860R
BOSSBS-12860REA 725 EUR
BOSSBS-12860R
Anti-CRYGS Rabbit Polyclonal Antibody
Antibodies
Crystallins are separated into two classes:taxon-specific, or enzyme, and ubiquitous. The latter classconstitutes the major proteins of vertebrate eye lens and maintainsthe transparency and refractive index of the lens. Since lenscentral fiber cells lose their nuclei during development, thesecrystallins are made and then retained throughout life, making themextremely stable proteins. Mammalian lens crystallins are dividedinto alpha, beta, and gamma families; beta and gamma crystallinsare also considered as a superfamily. Alpha and beta families arefurther divided into acidic and basic groups.More Product Information