Anti-Glycogenin 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Supplier: Bioss
BS-13447R-A680
BOSSBS-13447R-A680EA 890 EUR
BOSSBS-13447R-A680
Anti-Glycogenin 1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Glycogen synthesis is initiated by the autoglucosylation of Glycogenin-1. Specifically, Glycogenin-1 glucosylates itself to begin the synthesis of glycogen in mammalian skeletal muscle. It acts as the primer to which further glucose monomers may be added. All of the Glycogenin-1 molecules contain at least one glucosyl residue before autoglucosylation begins. The first step of the glycogen synthesis occurs when a glucose molecule from UDP-glucose binds to the hydroxyl group of Tyr 194 on the Glycogenin-1 molecule. Using its glucosyltransferase activity, Glycogenin-1 adds more glucoses, eachMore Product Information