Anti-GMEB2 Rabbit Polyclonal Antibody (Cy5®)
Supplier: Bioss
BS-13456R-CY5
BOSSBS-13456R-CY5EA 900 EUR
BOSSBS-13456R-CY5
Anti-GMEB2 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
GMEB-2 is a DNA-binding protein that plays a crucial role modulating transcription upon activation by steroid hormones. GMEB-2 is ubiquitously expressed with preferential expression in developmentally important tissues, such as testis, bone marrow, placenta and fetal tissues. It localizes to the nucleus and cytoplasm and contains a SAND domain near its N-terminus and a C-terminal coiled coil structure. GMEB-2 functions as a homodimer or as a heterodimer with GMEB-1. The formed complex specifically binds to glucocorticoid modulatory elements (GME) in the promoter region of target genes andMore Product Information