Anti-GOLGA7 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Supplier: Bioss
BS-13484R-A680
BOSSBS-13484R-A680EA 890 EUR
BOSSBS-13484R-A680
Anti-GOLGA7 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
GOLGA7 is a multi-pass membrane protein belonging to the Erf4 family of proteins. It is the functional ortholog of the yeast Erf4 protein. Localizing to the Golgi apparatus, GOLGA7 is a widely expressed protein but its expression is absent from colon and thymus tissues. GOLGA7 is palmitoylated on two cysteine residues, and this palmitoylation is required for its interaction with golgin 160 and its Golgi-localization. GOLGA7 also forms a complex with ZDHHC9 and, together, these proteins function as a Ras palmitoyltransferase (Ras PAT) which is required for palmitoylation of H-Ras and N-RasMore Product Information