Anti-PSMD9 Rabbit Polyclonal Antibody (Cy5.5®)
Supplier: Bioss
BS-1372R-CY5.5
BOSSBS-1372R-CY5.5EA 900 EUR
BOSSBS-1372R-CY5.5
Anti-PSMD9 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
The 26S proteasome is a multicatalytic proteinase complex with a highly ordered structure composed of 2 complexes, a 20S core and a 19S regulator. The 20S core is composed of 4 rings of 28 non-identical subunits; 2 rings are composed of 7 alpha subunits and 2 rings are composed of 7 beta subunits. The 19S regulator is composed of a base, which contains 6 ATPase subunits and 2 non-ATPase subunits, and a lid, which contains up to 10 non-ATPase subunits. Proteasomes are distributed throughout eukaryotic cells at a high concentration and cleave peptides in an ATP/ubiquitin-dependent process in aMore Product Information