Anti-Hck Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-1438R-CY5.5
BOSSBS-1438R-CY5.5EA 900 EUR
BOSSBS-1438R-CY5.5
Anti-Hck Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
HCK is a tyrosine kinase that is predominantly expressed in hemopoietic cell types. It may help couple the Fc receptor to the activation of the respiratory burst. In addition, it may play a role in neutrophil migration and in the degranulation of neutrophils.

Type: Primary
Antigen: Hck
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5
Public Immunogen Range: 301-450/526
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR