Anti-A2AR/Adenosine A2a Receptor Rabbit Polyclonal Antibody (HRP (Horseradish PE (Phycoerythrin)rOxidase))

Supplier: Bioss
BS-1456R-HRP
BOSSBS-1456R-HRPEA 883 EUR
BOSSBS-1456R-HRP
Anti-A2AR/Adenosine A2a Receptor Rabbit Polyclonal Antibody (HRP (Horseradish PE (Phycoerythrin)rOxidase))
Antibodies
This gene encodes a protein which is one of several receptor subtypes for adenosine. The activity of the encoded protein, a G protein coupled receptor family member, is mediated by G proteins which activate adenylyl cyclase. The encoded protein is abundant in basal ganglia, vasculature and platelets and it is a major target of caffeine.

Type: Primary
Antigen: A2AR/Adenosine A2a receptor
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 165-300/412
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name A2AR/Adenosine A2a receptor
Antigen synonyms A2aR|ADORA2|ADORA2A|Adenosine receptor A2a|RDC8
Antibody type Primary
Clonality Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 135
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
Modification Unmodified
Epitope 165-300/412
Immunogen KLH conjugated synthetic peptide derived from human A2a receptor
Form Liquid
Molecular weight 45 kDa
Purification Purified by Protein A
Concentration 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Storage temperature Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR