Anti-AQP1 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-1506R-CY5.5
BOSSBS-1506R-CY5.5EA 909 EUR
BOSSBS-1506R-CY5.5
Anti-AQP1 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Forms a water-specific channel that provides the plasma membranes of red cells and kidney proximal tubules with high permeability to water, thereby permitting water to move in the direction of an osmotic gradient.

Type: Primary
Antigen: AQP1
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5
Public Immunogen Range: 181-269/269
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR