Anti-C20orf70 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss
BS-15114R-FITC
BOSSBS-15114R-FITCEA 900 EUR
BOSSBS-15114R-FITC
Anti-C20orf70 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
C20orf70

Type: Primary
Antigen: C20orf70
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 151-249/249
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR