Anti-C2orf76 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-15160R-CY7
BOSSBS-15160R-CY7EA 900 EUR
BOSSBS-15160R-CY7
Anti-C2orf76 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
C2orf76

Type: Primary
Antigen: C2orf76
Clonality: Polyclonal
Clone:
Conjugation: Cy7
Public Immunogen Range: 1-80/126
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR