Anti-mucin3/Muc3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
BS-1525R-A750
BOSSBS-1525R-A750EA 899 EUR
BOSSBS-1525R-A750
Anti-mucin3/Muc3 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Major glycoprotein component of a variety of mucus gels. Thought to provide a protective, lubricating barrier against particles and infectious agents at mucosal surfaces. May be involved in ligand binding and intracellular signaling.

Type: Primary
Antigen: mucin3/Muc3
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1501-1600/1719
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name mucin3/Muc3
Antigen synonyms MUC3A|MUC-3A|MUC3|Intestinal mucin-3A|Mucin-3A
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 750
Reactivity Human
Host Rabbit
Gene ID 4584
Isotype IgG
ImmunoChemistry Yes
ImmunoFluorescence Yes
Modification Unmodified
Epitope 1501-1600/1719
Immunogen KLH conjugated synthetic peptide derived from human mucin3
Form Liquid
Molecular weight 264 kDa
Purification Purified by Protein A
Concentration 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Storage temperature Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR