Anti-Heparanase Rabbit Polyclonal Antibody (Cy5®)
Supplier: Bioss
BS-1541R-CY5
BOSSBS-1541R-CY5EA 876 EUR
BOSSBS-1541R-CY5
Anti-Heparanase Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Endoglycosidase that cleaves heparan sulfate proteoglycans (HSPGs) into heparan sulfate side chains and core proteoglycans. Participates in extracellular matrix (ECM) degradation and remodelling. Selectively cleaves the linkage between a glucuronic acid unit and an N-sulfo glucosamine unit carrying either a 3-O-sulfo or a 6-O-sulfo group. Can also cleave the linkage between a glucuronic acid unit and an N-sulfo glucosamine unit carrying a 2-O-sulfo group, but not linkages between a glucuronic acid unit and a 2-O-sulfated iduronic acid moiety. It is essentially inactive at neutral pH butMore Product Information