Anti-HGD Rabbit Polyclonal Antibody (FITC)
Supplier: Bioss
BS-15472R-FITC
BOSSBS-15472R-FITCEA 900 EUR
BOSSBS-15472R-FITC
Anti-HGD Rabbit Polyclonal Antibody (FITC)
Antibodies
HGD is a 445 amino acid protein that belongs to the homogentisate dioxygenase family and is involved in the pathway of amino acid degradation. Expressed at high levels in kidney, colon, liver, prostate and small intestine, HGD uses iron as a cofactor to catalyse the oxygen-dependent conversion of homogentisate to 4-maleylacetoacetate, a reaction that is the fourth step in the creation of L-phenylalanine from fumarate and acetoacetic acid. Defects in the gene encoding HGD are the cause of alkaptonuria (AKU), an autosomal recessive disorder that is characterised by urine that turns dark onMore Product Information