Anti-HIBCH Rabbit Polyclonal Antibody (Cy3)
Supplier: Bioss
BS-15484R-CY3
BOSSBS-15484R-CY3EA 900 EUR
BOSSBS-15484R-CY3
Anti-HIBCH Rabbit Polyclonal Antibody (Cy3)
Antibodies
HIBCH is a 386 amino acid protein belonging to the enoyl-CoA hydratase/isomerase family. Localising to the mitochondria, HIBCH is highly expressed in liver and kidney, with lower levels found in heart, muscle and brain. HIBCH hydrolyses HIBYL-CoA, a saline catabolite, and _-hydroxypropionyl-CoA, an intermediate in the minor pathway involved in the metabolism of proprionate. Existing as two alternatively spliced isoforms, the gene encoding HIBCH maps to human chromosome 2q32.2. Defects to this gene result in HIBCH deficiency (HIBCHD), known alternatively as deficiency of _-hydroxyisobutyrylMore Product Information