Anti-FHIT Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Supplier: Bioss
BS-1769R-A350
BOSSBS-1769R-A350EA 901 EUR
BOSSBS-1769R-A350
Anti-FHIT Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Cleaves P(1)-P(3)-bis(5'-adenosyl) triphosphate (Ap3A) to yield AMP and ADP. Can also hydrolyze P(1)-P(4)-bis(5'-adenosyl) tetraphosphate (Ap4A), but has extremely low activity with ATP. Modulates transcriptional activation by CTNNB1 and thereby contributes to regulate the expression of genes essential for cell proliferation and survival, such as CCND1 and BIRC5. Plays a role in the induction of apoptosis via SRC and AKT1 signaling pathways. Inhibits MDM2-mediated proteasomal degradation of p53/TP53 and thereby plays a role in p53/TP53-mediated apoptosis. Induction of apoptosis depends onMore Product Information