Anti-SLC25A26 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)

Supplier: Bioss
BS-19813R-A680
BOSSBS-19813R-A680EA 890 EUR
BOSSBS-19813R-A680
Anti-SLC25A26 Rabbit Polyclonal Antibody (ALEXA FLUOR® 680)
Antibodies
Mitochondrial solute carriers shuttle metabolites, nucleotides, and cofactors through the mitochondrial inner membrane. Specifically mediates the transport of S-adenosylmethionine (SAM) into the mitochondria.

Type: Primary
Antigen: SLC25A26
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/274
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR