Anti-AIMP2 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-2509R-CY5
BOSSBS-2509R-CY5EA 909 EUR
BOSSBS-2509R-CY5
Anti-AIMP2 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Required for assembly and stability of the aminoacyl-tRNA synthase complex. Mediates ubiquitination and degradation of FUBP1, a transcriptional activator of MYC, leading to MYC down-regulation which is required for aveolar type II cell differentiation. Blocks MDM2-mediated ubiquitination and degradation of p53/TP53. Functions as a proapoptotic factor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: AIMP2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR