Anti-IL-21 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
BS-2621R-A750
BOSSBS-2621R-A750EA 899 EUR
BOSSBS-2621R-A750
Anti-IL-21 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Cytokine with immunoregulatory activity. May promote the transition between innate and adaptive immunity. Induces the production of IgG(1) and IgG(3) in B-cells (By similarity). May play a role in proliferation and maturation of natural killer (NK) cells in synergy with IL15. May regulate proliferation of mature B- and T-cells in response to activating stimuli. In synergy with IL15 and IL18 stimulates interferon gamma production in T-cells and NK cells. During T-cell mediated immune response may inhibit dendritic cells (DC) activation and maturation.

Type: Primary
Antigen: IL-21
Clonality:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR