Anti-ENDOG Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-2800R-A488
BOSSBS-2800R-A488EA 893 EUR
BOSSBS-2800R-A488
Anti-ENDOG Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
The protein encoded by this gene is a nuclear encoded endonuclease that is localized in the mitochondrion. The encoded protein is widely distributed among animals and cleaves DNA at GC tracts. This protein is capable of generating the RNA primers required by DNA polymerase gamma to initiate replication of mitochondrial DNA. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ENDOG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol ENDOG
Environmentally Preferable
Antigen name Endonuclease G, mitochondrial
Antigen synonyms endonuclease G|mitochondrial endonuclease G|endo G|mitochondrial
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 488
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 362100
Isotype IgG
ImmunoChemistry Yes
Immunogen 110-150/297
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR