Anti-VDR Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-2987R-A555
BOSSBS-2987R-A555EA 901 EUR
BOSSBS-2987R-A555
Anti-VDR Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Nuclear hormone receptor. Transcription factor that mediates the action of vitamin D3 by controlling the expression of hormone sensitive genes. Regulates transcription of hormone sensitive genes via its association with the WINAC complex, a chromatin-remodeling complex. Recruited to promoters via its interaction with the WINAC complex subunit BAZ1B/WSTF, which mediates the interaction with acetylated histones, an essential step for VDR-promoter association. Plays a central role in calcium homeostasis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: VDR
Clonality:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR