Anti-ALOX5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-3251R-HRP
BOSSBS-3251R-HRPEA 929 EUR
BOSSBS-3251R-HRP
Anti-ALOX5 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
This gene encodes a member of the lipoxygenase gene family and plays a dual role in the synthesis of leukotrienes from arachidonic acid. The encoded protein, which is expressed specifically in bone marrow-derived cells, catalyzes the conversion of arachidonic acid to 5(S)-hydroperoxy-6-trans-8,11,14-cis-eicosatetraenoic acid, and further to the allylic epoxide 5(S)-trans-7,9-trans-11,14-cis-eicosatetrenoic acid (leukotriene A4). Leukotrienes are important mediators of a number of inflammatory and allergic conditions. Mutations in the promoter region of this gene lead to a diminished responseMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol ALOX5
Antigen name Arachidonate 5-Lipoxygenase
Antigen synonyms arachidonic acid 5-lipoxygenase|5-LOX|arachidonic 5-lipoxygenase delta-10-13 isoform|leukotriene A4 synthase|5LPG|arachidonic 5-lipoxygenase alpha-10 isoform|5-LO|LOG5|arachidonic 5-lipoxygenase delta-13 isoform|arachidonic 5-lipoxygenase delta-p10 isoform
Antibody type Primary
Clonality Polyclonal
Conjugation HRP (Horseradish Peroxidase)
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 240
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Modification phospho-Ser271
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% Gentamicin. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR