Anti-PIN1 Rabbit Polyclonal Antibody (Cy5®)
Supplier: Bioss
BS-3354R-CY5
BOSSBS-3354R-CY5EA 1350 EUR
BOSSBS-3354R-CY5
Anti-PIN1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Essential PPIase that regulates mitosis presumably by interacting with NIMA and attenuating its mitosis-promoting activity. Displays a preference for an acidic residue N-terminal to the isomerized proline bond. Catalyzes pSer/Thr-Pro cis/trans isomerizations. Down-regulates kinase activity of BTK. Can transactivate multiple oncogenes and induce centrosome amplification, chromosome instability and cell transformation. Required for the efficient dephosphorylation and recycling of RAF1 after mitogen activation. Binds and targets PML and BCL6 for degradation in a phosphorylation-dependent manner.More Product Information