Anti-CHRDL2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Supplier: Bioss
BS-3860R-FITC
BOSSBS-3860R-FITCEA 900 EUR
BOSSBS-3860R-FITC
Anti-CHRDL2 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
CHRDL2 is a novel chordin like protein that can act as a BMP antagonist. Amember of the chordin family of proteins, it contains a signal peptide andthree CR (cysteine-rich repeat) domains. When expressed as a recombinantprotein it is secreted and binds to activin A, but not to BMP-2, -4, -6. Differential expression has been detected in developing chondrocytes, myoblasts, osteoblasts, and osteoarthritic joints. Complex alternative splicing of CHRDL2 potentially results in distinct isoforms that differ at their C termini, in the expression of signal peptide, and in the content of CR domains.More Product Information