Anti-Rab24 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
BS-3868R-A750
BOSSBS-3868R-A750EA 890 EUR
BOSSBS-3868R-A750
Anti-Rab24 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
May be involved in autophagy-related processes.

Type: Primary
Antigen: Rab24
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 1-100/203
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR