Anti-PTGES3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-4020R-A488
BOSSBS-4020R-A488EA 921 EUR
BOSSBS-4020R-A488
Anti-PTGES3 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Molecular chaperone that localizes to genomic response elements in a hormone-dependent manner and disrupts receptor-mediated transcriptional activation, by promoting disassembly of transcriptional regulatory complexes. Belongs to the p23/wos2 family.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PTGES3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR