Anti-MAT1A Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-4054R-A350
BOSSBS-4054R-A350EA 876 EUR
BOSSBS-4054R-A350
Anti-MAT1A Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
S adenosylmethionine synthetase catalyzes the formation of S adenosylmethionine from methionine and ATP. In mammalian tissues, there are three distinct forms of AdoMet synthases designated as alpha, beta, and gamma. Alpha and beta are expressed only in adult liver, while gamma is widely distributed in extrahepatic tissues.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAT1A
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR