Anti-CUL1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss
BS-4208R-HRP
BOSSBS-4208R-HRPEA 900 EUR
BOSSBS-4208R-HRP
Anti-CUL1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Cullin 1 may be involved in the degradation of positive cell cycle regulators (for example G1 cyclins in yeast). Human cullin 1 forms a complex with Skp1 and the F box protein Skp2, which is thought to play a role in the ubiquitination of G1 regulatory proteins, as its homolog does in yeast.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: CUL1
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR