Anti-ADORA1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-4235R-CY5
BOSSBS-4235R-CY5EA 900 EUR
BOSSBS-4235R-CY5
Anti-ADORA1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Receptor for adenosine. The activity of this receptor is mediated by G proteins which inhibit adenylyl cyclase.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ADORA1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

SPECIFICATIONS

Antigen symbol ADORA1
Environmentally Preferable
Antigen name Adenosine A1 Receptor
Antigen synonyms RDC7
Antibody type Primary
Clonality Polyclonal
Conjugation Cy5®
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 134
Isotype IgG
ImmunoChemistry Yes
Immunogen 147-176/326
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR