Anti-ICAM1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-4618R-A555
BOSSBS-4618R-A555EA 893 EUR
BOSSBS-4618R-A555
Anti-ICAM1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
ICAM proteins are ligands for the leukocyte adhesion protein LFA-1 (integrin alpha-L/beta-2). During leukocyte trans-endothelial migration, ICAM1 engagement promotes the assembly of endothelial apical cups through ARHGEF26/SGEF and RHOG activation. In case of rhinovirus infection acts as a cellular receptor for the virus.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ICAM1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR