Anti-CYP39A1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-5050R-A350
BOSSBS-5050R-A350EA 893 EUR
BOSSBS-5050R-A350
Anti-CYP39A1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
The CYP39A1 gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of steroids, cholesterol and other lipids. This endoplasmic reticulum protein is involved in the conversion of cholesterol to bile acids. Its substrates include the oxysterols 25-hydroxycholesterol, 27-hydroxycholesterol and 24-hydroxycholesterol.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CYP39A1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR