Anti-CYP4A11 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
BS-5054R-A488
BOSSBS-5054R-A488EA 1350 EUR
BOSSBS-5054R-A488
Anti-CYP4A11 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and hydroxylates medium-chain fatty acids such as laurate and myristate. [provided by RefSeq, Jul 2008].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CYP4A11
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR