Anti-ATF2 Rabbit Polyclonal Antibody
Supplier: Bioss
BS-5170R
BOSSBS-5170REA 743 EUR
BOSSBS-5170R
Anti-ATF2 Rabbit Polyclonal Antibody
Antibodies
ATF2 is a member of the ATF/CREB family of basic region leucine zipper DNA binding proteins that regulates transcription by binding to a consensus cAMP response element (CRE) in the promoter of various viral and cellular genes. Many of these genes are important in cell growth and differentiation, and in stress and immune responses. ATF2 is a nuclear protein that binds DNA as a dimer and can form dimers with members of the ATF/CREB and Jun/Fos families. It is a stronger activator as a heterodimer with cJun than as a homodimer. Several isoforms of ATF2 arise by differential splicing. TheMore Product Information