Anti-DIO3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-5331R-A647
BOSSBS-5331R-A647EA 922 EUR
BOSSBS-5331R-A647
Anti-DIO3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
mTOR is one of a family of proteins involved in cell cycle progression, DNA recombination, and DNA damage detection. In rat, it is a 289-kDa protein (symbolized RAFT1) with significant homology to the Saccharomyces cerevisiae protein TOR1 and has been shown to associate with the immunophilin FKBP12 in a rapamycin dependent fashion. The FKBP12-rapamycin complex is known to inhibit progression through the G1 cell cycle stage by interfering with mitogenic signaling pathways involved in G1 progression in several cell types, as well as in yeast. The binding of FRAP to FKBP12-rapamycin correlatedMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol DIO3
Environmentally Preferable
Antigen name deiodinase, iodothyronine, type III
Antigen synonyms TXDI3|DIOIII|D3|5DIII
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 647
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 1735
Isotype IgG
ImmunoChemistry Yes
Modification phospho-Thr2446
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at 4°C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR