Anti-EIF4B Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-5346R-CY5.5
BOSSBS-5346R-CY5.5EA 930 EUR
BOSSBS-5346R-CY5.5
Anti-EIF4B Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
eIF4B is required for the binding of mRNA to ribosomes. It functions in close association with eIF4F and eIF4A. It binds near the 5'-terminal cap of mRNA in the presence of eIF4F and ATP. It promotes the ATPase activity and the ATP-dependent RNA unwinding activity of both eIF4A and eIF4F (referenced from swissprot).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EIF4B
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR