Anti-RPS6KB1 Ser441+Thr444 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
BS-5673R-A680
BOSSBS-5673R-A680EA 921 EUR
BOSSBS-5673R-A680
Anti-RPS6KB1 Ser441+Thr444 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
This gene encodes a member of the ribosomal S6 kinase family of serine/threonine kinases. The encoded protein responds to mTOR (mammalian target of rapamycin) signaling to promote protein synthesis, cell growth, and cell proliferation. Activity of this gene has been associated with human cancer. Alternatively spliced transcript variants have been observed. The use of alternative translation start sites results in isoforms with longer or shorter N-termini which may differ in their subcellular localizations. There are two pseudogenes for this gene on chromosome 17.

Type: Primary
Antigen: RPS6More Product Information
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name RPS6KB1 Ser441+Thr444
Antigen synonyms p70S6K-alpha|Serine/threonine-protein kinase 14A|Ribosomal protein S6 kinase beta-1|70kDa|p70-S6K 1|PS6K|S6K-beta-1|p70 S6 Kinase|KS6B1_HUMAN|70 kDa ribosomal protein S6 kinase 1|ribosomal protein S6 kinase|P70S6K1|p70 S6K-alpha|RPS6KB1|RPS6KB1 phospho S441+T444|S6K|p70 S6 kinase alpha|p70 ribosomal S6 kinase alpha|S6K1|STK14A|Ribosomal protein S6 kinase I|p70 S6KA|P70 Beta1|p-RPS6KB1 Ser441+Thr444|p70-alpha|RPS6KB1 phospho Ser441+Thr444|p70-S6K
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 680
Reactivity Rat,Mouse
Host Rabbit
Gene ID 6198
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
ImmunoFluorescence Yes
Modification Phosphorylation (Ser441+Thr444)
Immunogen KLH conjugated synthetic phosphopeptide derived from human RPS6KB1 around the phosphorylation site of Ser441+Thr444
Form Liquid
Molecular weight 58 kDa
Purification Purified by Protein A
Concentration 1 µg/µl
Storage buffer Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
Storage temperature Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
Shipping temperature 4 °C
Tested applications ICC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR