Anti-KCNK1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Supplier: Bioss
BS-5933R-A555
BOSSBS-5933R-A555EA 893 EUR
BOSSBS-5933R-A555
Anti-KCNK1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
KCNK9 or TASK-3 (TWIK-related Acid sensitive K+ channel) is a member of the potassium channel family of proteins that contain two-pore domain and four transmembrane domains. These channels are characterized as leak K+ channels that are sensitive to changes in the extracellular pH. The physiological functions of TASK channels are largely unknown; it has been proposed that they may be involved in the regulation of breathing, aldosterone secretion and anesthetic-mediated neuronal activity. They were found to act in neurons' membrane potential and in resting K+ currents.
Recommended Dilutions:More Product Information