Anti-CST7 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Supplier: Bioss
BS-6039R-A488
BOSSBS-6039R-A488EA 893 EUR
BOSSBS-6039R-A488
Anti-CST7 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
The cystatin superfamily encompasses proteins that contain multiple cystatin-like sequences. Some of the members are active cysteine protease inhibitors, while others have lost or perhaps never acquired this inhibitory activity. There are three inhibitory families in the superfamily, including the type 1 cystatins (stefins), type 2 cystatins and the kininogens. The type 2 cystatin proteins are a class of cysteine proteinase inhibitors found in a variety of human fluids and secretions. Expression of the protein has been observed in various human cancer cell lines established from malignantMore Product Information