Anti-HPGD Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
BS-6051R-A350
BOSSBS-6051R-A350EA 915 EUR
BOSSBS-6051R-A350
Anti-HPGD Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Prostaglandin inactivation. Contributes to the regulation of events that are under the control of prostaglandin levels. Catalyzes the NAD-dependent dehydrogenation of lipoxin A4 to form 15-oxo-lipoxin A4. Inhibits in vivo proliferation of colon cancer cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HPGD
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR