Anti-MFSD2A Rabbit Polyclonal Antibody (Cy3®)
Supplier: Bioss
BS-6073R-CY3
BOSSBS-6073R-CY3EA 897 EUR
BOSSBS-6073R-CY3
Anti-MFSD2A Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Sodium-dependent lysophosphatidylcholine (LPC) symporter, which plays an essential role for blood-brain barrier formation and function. Specifically expressed in endothelium of the blood-brain barrier of micro-vessels and transports LPC into the brain. Transport of LPC is essential because it constitutes the major mechanism by which docosahexaenoic acid (DHA), an omega-3 fatty acid that is essential for normal brain growth and cognitive function, enters the brain. Transports LPC carrying long-chain fatty acids such LPC oleate and LPC palmitate with a minimum acyl chain length of 14 carbons.More Product Information