Anti-RING1 Rabbit Polyclonal Antibody

Supplier: Bioss
BS-6142R
BOSSBS-6142REA 725 EUR
BOSSBS-6142R
Anti-RING1 Rabbit Polyclonal Antibody
Antibodies
This gene belongs to the RING finger family, members of which encode proteins characterized by a RING domain, a zinc-binding motif related to the zinc finger domain. The gene product can bind DNA and can act as a transcriptional repressor. It is associated with the multimeric polycomb group protein complex. The gene product interacts with the polycomb group proteins BMI1, EDR1, and CBX4, and colocalizes with these proteins in large nuclear domains. It interacts with the CBX4 protein via its glycine-rich C-terminal domain. The gene maps to the HLA class II region, where it is contiguous withMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol RING1
Environmentally Preferable
Antigen name Ring Finger Protein 1
Antigen synonyms RING1A |OTTHUMP00000029416|RNF1|EC 6.3.2.-
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 6015
Isotype IgG
Western blot Yes
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A.
Concentration 1 ug/ul
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
Shipping temperature 4 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR