Anti-PCMT1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-6211R-CY5
BOSSBS-6211R-CY5EA 900 EUR
BOSSBS-6211R-CY5
Anti-PCMT1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Catalyzes the methyl esterification of L-isoaspartyl and D-aspartyl residues in peptides and proteins that result from spontaneous decomposition of normal L-aspartyl and L-asparaginyl residues. It plays a role in the repair and/or degradation of damaged proteins. Acts on EIF4EBP2, microtubule-associated protein 2, calreticulin, clathrin light chains a and b, Ubiquitin carboxyl-terminal hydrolase isozyme L1, phosphatidylethanolamine-binding protein 1, stathmin, beta-synuclein and alpha-synuclein.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PCMT1
Clonality: Polyclonal
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR