Anti-RAB25 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
BS-6501R-CY7
BOSSBS-6501R-CY7EA 900 EUR
BOSSBS-6501R-CY7
Anti-RAB25 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
RAB proteins, such as RAB25, are members of the RASsuperfamily of small GTPases that are involved in membranetrafficking. Members of the RAB11 subfamily, including RAB25,control the return of internalized membrane-associated moieties tothe cell surface

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RAB25
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR